Tetrabutylammonium hydroxide 30-hydrate, 95+%
Tetrabutylammonium hydroxide 30-hydrate, 95+%
Tetrabutylammonium hydroxide 30-hydrate, 95+%
Tetrabutylammonium hydroxide 30-hydrate, 95+%
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide 30-hydrate, 95+%

CAS: 147741-30-8 | C16H97NO31 | 799.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 434830250
Price (USD)
101.00
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
101.00
Each
Add to cart
Chemical Identifiers
CAS147741-30-8
IUPAC Nametetrabutylazanium triacontahydrate hydroxide
Molecular FormulaC16H97NO31
InChI KeyDFGIRVKFXSRUOX-UHFFFAOYSA-M
SMILESO.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl95.0 to 105.0 %
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Low melting fused mass or liquid
Appearance (Form)Powder or crystals and/or chunks or
Melting point26°C to 30°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link