Thermo Scientific Chemicals

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

Catalog number: 434831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

Catalog number: 434831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 434831000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147741-30-8
IUPAC Name
tetrabutylazanium triacontahydrate hydroxide
Molecular Formula
C16H97NO31
InChI Key
DFGIRVKFXSRUOX-UHFFFAOYSA-M
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Titration with HCl
95.0 to 105.0 %
Appearance (Color)
Colorless to white to light yellow
Appearance (Form)
Low melting fused mass or liquid
Appearance (Form)
Powder or crystals and/or chunks or
Melting point
26°C to 30°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text