BOC-L-Serine benzyl ester, 98%, Thermo Scientific Chemicals
BOC-L-Serine benzyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-Serine benzyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 434840050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS59524-02-6
IUPAC Namebenzyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Molecular FormulaC15H21NO5
InChI KeyBQADRZHPZVQGCW-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Melting point65°C to 75°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only