O-tert-Butyl-L-serine, 97%, Thermo Scientific Chemicals
O-tert-Butyl-L-serine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

O-tert-Butyl-L-serine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 434880050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-amino-3-(tert-butoxy)propanoic acid
Molecular FormulaC7H15NO3
InChI KeyDDCPKNYKNWXULB-UHFFFAOYNA-N
SMILESCC(C)(C)OCC(N)C(O)=O
Molecular Weight (g/mol)161.20
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-15.5° to -17.5° (20°C, 589 nm)
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only