Thermo Scientific Chemicals

Trioctylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 434900250
25 g, Each
Thermo Scientific Chemicals

Trioctylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 434900250
25 g, Each
Quantity
Catalog number: 434900250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18765-09-8
IUPAC Name
trioctylsilyl
Molecular Formula
C24H51Si
InChI Key
QTKHQYWRGFZFHG-UHFFFAOYSA-N
SMILES
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94.0 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text