4-Ethoxybenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Ethoxybenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethoxybenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 434970010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS16331-46-7
IUPAC Name4-ethoxybenzoyl chloride
Molecular FormulaC9H9ClO2
InChI KeyXLWQUESMILVIPR-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only