1,3-Dicyclohexylbenzimidazolium chloride, 95%, Thermo Scientific Chemicals
1,3-Dicyclohexylbenzimidazolium chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dicyclohexylbenzimidazolium chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435095000
Price (USD)/ Each
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS1034449-15-4
IUPAC Name1,3-dicyclohexyl-3H-1λ⁵,3-benzodiazol-1-ylium chloride
Molecular FormulaC19H27ClN2
InChI KeyJACUDTKAZHCIPU-UHFFFAOYSA-M
SMILES[Cl-].C1CCC(CC1)N1C=[N+](C2CCCCC2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis67.30 to 75.90 % (C)
Elemental Analysis8.25 to 9.31 % (N)
Appearance (Color)White to grey to tan
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only