Applied Biosystems™

7500 Real Time PCR Systems Spectral Calibration Kit II

Catalog number: 4351151
Have Questions?
Applied Biosystems™

7500 Real Time PCR Systems Spectral Calibration Kit II

Catalog number: 4351151
Have Questions?
Catalog Number
4351151
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43511511 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This easy-to-use kit enables you to establish the pure dye spectra and multicomponent values for Cy3®, Cy5®, and Texas Red™ dyes on the 7500 Real-Time PCR System. The Applied Biosystems® 7500 Real-Time PCR Systems Spectral Calibration Kit II contains three optical 96-well reaction plates preloaded and sealed with Cy3®, Cy5®, and Texas Red™ fluorescent dye standards.

• 96-well optical reaction plates preloaded with Cy3®, Cy5®, and Texas Red™ dyes allow quick and easy calibration of the 7500 Real-Time PCR System
• Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously

Spend Your Time on Research, Not Instrument Calibration
This kit is easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 7500 Real-Time PCR System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibrated Dye
Cy™3, Cy™5, Texas Red™
For Use With (Equipment)
7500 System
Detection Method
Fluorescent
Product Type
Spectral Calibration Kit
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Dry Ice
Calibration Type
Spectral Calibration

Contents & Storage

Contains three Optical 96-Well Reaction Plates, preloaded and sealed containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards.

Store at -15 to -25°C. (This product is light sensitive.)

Expiration date: 12 months from the date of delivery

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text