Chemical Identifiers
CAS213343-64-7
IUPAC Nameλ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl}-2,5-diethylphospholane tetrafluoroboranuide
Molecular FormulaC30H48BF4P2Rh
InChI KeyDSYBEQKPSKLNMC-RTXTXWNLNA-N
SMILES[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@H]1CC[C@H](CC)P1C1=C(C=CC=C1)P1[C@@H](CC)CC[C@@H]1CC
View more