Chemical Identifiers
CAS213343-65-8
IUPAC Nameλ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-dimethylphospholan-1-yl]ethyl}-2,5-dimethylphospholane tetrafluoroboranuide
Molecular FormulaC22H40BF4P2Rh
InChI KeyCVUPEPGZCZHSOF-BDNLEJQTNA-N
SMILES[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C[C@H]1CC[C@H](C)P1CCP1[C@@H](C)CC[C@@H]1C
View more