Thermo Scientific Chemicals

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 435320250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 435320250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435320250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14316-06-4
IUPAC Name
hydrogen methyl 2-aminopropanoate chloride
Molecular Formula
C4H10ClNO2
InChI Key
IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(C)N
Appearance (Color)
White
Titration Argentometric
>=97.5 to =<102.5 %
Infrared spectrum
Conforms
Appearance (Form)
Powder or solid
Specific optical rotation
-10° to -15° (20°C, 589 nm) c=2, methanol

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text