Thermo Scientific Chemicals

4-Fluoro-3-methylphenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 435330050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Fluoro-3-methylphenylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 435330050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435330050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139911-27-6
IUPAC Name
(4-fluoro-3-methylphenyl)boronic acid
Molecular Formula
C7H8BFO2
InChI Key
JCIJCHSRVPSOML-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1F)B(O)O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text