4-Fluoro-3-methylphenylboronic acid, 98%
4-Fluoro-3-methylphenylboronic acid, 98%
4-Fluoro-3-methylphenylboronic acid, 98%
4-Fluoro-3-methylphenylboronic acid, 98%
Thermo Scientific Chemicals

4-Fluoro-3-methylphenylboronic acid, 98%

CAS: 139911-27-6 | C7H8BFO2 | 153.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 435330250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS139911-27-6
IUPAC Name(4-fluoro-3-methylphenyl)boronic acid
Molecular FormulaC7H8BFO2
InChI KeyJCIJCHSRVPSOML-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1F)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection