BOC-L-Leucine monohydrate, 99%, Thermo Scientific Chemicals
BOC-L-Leucine monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-Leucine monohydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 435490050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS200936-87-4
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid hydrate
Molecular FormulaC11H23NO5
InChI KeyURQQEIOTRWJXBA-UHFFFAOYNA-N
SMILESO.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point85°C to 90°C
Specific optical rotation-22.5° to -27.5° (20°C, 589 nm) (c=2, glacial acetic acid)
HPLC>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only