Thermo Scientific Chemicals

BOC-L-Leucine monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 435490250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

BOC-L-Leucine monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 435490250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435490250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
200936-87-4
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid hydrate
Molecular Formula
C11H23NO5
InChI Key
URQQEIOTRWJXBA-UHFFFAOYNA-N
SMILES
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
Melting point
85°C to 90°C
Specific optical rotation
-22.5° to -27.5° (20°C, 589 nm) (c=2, glacial acetic acid)
HPLC
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text