Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435558000
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
Add to cart
Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Catalog number435558000
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
336.00 
Save 50.35 (15%)
-
Add to cart
Chemical Identifiers
CAS693-25-4
IUPAC Namebromo(pentyl)magnesium
Molecular FormulaC5H11BrMg
InChI KeyZXWGTPYWPZGZPT-UHFFFAOYSA-M
SMILESCCCCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Yellow to brown to black
Titration Grignard1.90 to 2.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only