Thermo Scientific Chemicals

(S)-2-Amino-7-hydroxytetralin, 95%, Thermo Scientific Chemicals

Catalog number: 435590010
1 g, Each
Thermo Scientific Chemicals

(S)-2-Amino-7-hydroxytetralin, 95%, Thermo Scientific Chemicals

Catalog number: 435590010
1 g, Each
Quantity
Catalog number: 435590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85951-60-6
IUPAC Name
7-amino-5,6,7,8-tetrahydronaphthalen-2-ol
Molecular Formula
C10H13NO
InChI Key
VIYAPIMIOKKYNF-UHFFFAOYNA-N
SMILES
NC1CCC2=C(C1)C=C(O)C=C2
Appearance (Form)
Crystalline Powder or crystals
Melting point
144°C to 151°C
Appearance (Color)
White to grey
Titration with HClO4
94 to 106 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text