Thermo Scientific Chemicals

Butyltrichlorosilane, 99%, Thermo Scientific Chemicals

Catalog number: 435600250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Butyltrichlorosilane, 99%, Thermo Scientific Chemicals

Catalog number: 435600250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 435600250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7521-80-4
IUPAC Name
butyltrichlorosilane
Molecular Formula
C4H9Cl3Si
InChI Key
FQEKAFQSVPLXON-UHFFFAOYSA-N
SMILES
CCCC[Si](Cl)(Cl)Cl
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4360 to 1.4380 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text