n-Nonane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
n-Nonane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Nonane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435631000
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
101.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
101.00 
Save 15.35 (15%)
Add to cart
n-Nonane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Catalog number435631000
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
101.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
CAS111-84-2
IUPAC Namenonane
Molecular FormulaC9H20
InChI KeyBKIMMITUMNQMOS-UHFFFAOYSA-N
SMILESCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Water=<0.005 %
Color scale=<10 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only