5-Iodo-1H-indazole, 95%, Thermo Scientific Chemicals
5-Iodo-1H-indazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Iodo-1H-indazole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435710050
Price (USD)/ Each
659.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
776.00 
Save 116.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
659.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
776.00 
Save 116.35 (15%)
Add to cart
5-Iodo-1H-indazole, 95%, Thermo Scientific Chemicals
Catalog number435710050
Price (USD)/ Each
659.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
776.00 
Save 116.35 (15%)
-
Add to cart
Chemical Identifiers
CAS55919-82-9
IUPAC Name5-iodo-1H-indazole
Molecular FormulaC7H5IN2
InChI KeyCGCHCLICSHIAAM-UHFFFAOYSA-N
SMILESIC1=CC=C2NN=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point151°C to 156°C
Appearance (Color)Yellow to brown to purple
Infrared spectrumConforms
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only