Thermo Scientific Chemicals

(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, Thermo Scientific Chemicals

Catalog number: 435830010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, Thermo Scientific Chemicals

Catalog number: 435830010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435830010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104530-79-2
IUPAC Name
oxolan-3-amine
Molecular Formula
C4H9NO
InChI Key
MIPHRQMEIYLZFZ-UHFFFAOYNA-N
SMILES
NC1CCOC1
Infrared spectrum
Conforms
Appearance (Form)
Crystals or solid
Appearance (Color)
White to off-white to light brown
Elemental Analysis
48.9 to 53.0 % C
Specific optical rotation
-3° to -5° (20°C, 589 nm) (c=1, methanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text