The Applied Biosystems™ MicroAmp™ Fast 96-Well Tray, specifically designed to snugly hold MicroAmp™ Fast Reaction Tubes with Cap (0.1 mL) and MicroAmp™ Fast 8-Tube Strips, is required for more efficient sample processing on the Applied Biosystems™ 9800 Fast Thermal Cycler.
• Organizes your Fast Reaction Tubes or Strips • Provides the thermal contact necessary for fast PCR • Manufactured to high specification, ensuring a quality product
Convenience and Performance The MicroAmp™ Fast 96-Well Tray allows you to organize your tubes while adding sample and mix. This handy tray reduces spillage and tube mix-ups by securing and ordering your tubes on the lab bench. When the samples are ready, simply put the tray of tubes directly on the block of the 9800 Fast Thermal Cycler. Not only does the tray eliminate the hassle of transferring the tubes, but it also provides the thermal contact necessary for efficient and accurate thermal cycling.
Specifications
Certifications/Compliance
Certified DNA/RNase Free
Product Line
MicroAmp
Product Type
Tray
For Use With (Equipment)
9800 Fast Thermal Cycler
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
10 Trays
Shipping Condition
Room Temperature
Contents & Storage
Contains 10 MicroAmp™ Fast 96-Well Trays. Store at room temperature.