Thermo Scientific Chemicals

Isoindoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 435850250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Isoindoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 435850250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435850250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32372-82-0
IUPAC Name
2,3-dihydro-1H-isoindole hydrochloride
Molecular Formula
C8H10ClN
InChI Key
NOVIRODZMIZUPA-UHFFFAOYSA-N
SMILES
Cl.C1NCC2=CC=CC=C12
Infrared spectrum
Conforms
Elemental Analysis
59.27 to 64.21 % C
Appearance (Form)
Powder
Elemental Analysis
8.64 to 9.36 % N
Appearance (Color)
White to off-white to grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text