Thermo Scientific Chemicals

2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 435920010
1 g, Each
Thermo Scientific Chemicals

2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 435920010
1 g, Each
Quantity
Catalog number: 435920010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
458532-94-0
IUPAC Name
(2-bromopyridin-4-yl)boronic acid
Molecular Formula
C5H5BBrNO2
InChI Key
DFIKFQMGZNTFLI-UHFFFAOYSA-N
SMILES
OB(O)C1=CC(Br)=NC=C1
Appearance
May have a pink hue
Appearance (Color)
White to brown
HPLC
>=94.0 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text