Thermo Scientific Chemicals

2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 435930050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 435930050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 435930050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
458532-86-0
IUPAC Name
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C11H15BFNO2
InChI Key
PCLMNCBIXQQRMB-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
GC
>=94.0 %
Appearance (Color)
Yellow to beige
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Melting point
47°C to 54°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text