Thermo Scientific Chemicals

Pentamethylenebis(magnesium bromide), 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

產品號碼: 436001000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Pentamethylenebis(magnesium bromide), 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

產品號碼: 436001000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
產品號碼: 436001000
價格 (TWD)
Have Questions?

化學識別

CAS
23708-48-7
IUPAC Name
bromo[5-(bromomagnesio)pentyl]magnesium
Molecular Formula
C5H10Br2Mg2
InChI Key
OUEGHZYBWJTUAD-UHFFFAOYSA-L
SMILES
Br[Mg]CCCCC[Mg]Br
Appearance (Color)
Yellow to grey to brown
Titration Grignard
0.45 to 0.65 M
Appearance (Form)
Solution

說明

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text