Thermo Scientific Chemicals

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436030500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436030500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 436030500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112022-81-8
IUPAC Name
(3aS)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular Formula
C18H20BNO
InChI Key
VMKAFJQFKBASMU-KRWDZBQOSA-N
SMILES
[H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
Strength
0.90 to 1.10 M
Appearance (Color)
Colorless to amber

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text