Thermo Scientific Chemicals

Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 436120050
5 g, Each
Thermo Scientific Chemicals

Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 436120050
5 g, Each
Quantity
Catalog number: 436120050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25602-68-0
IUPAC Name
8-azabicyclo[3.2.1]octan-3-one hydrochloride
Molecular Formula
C7H12ClNO
InChI Key
MZQWQFWRSDNBPV-UHFFFAOYNA-N
SMILES
Cl.O=C1CC2CCC(C1)N2
Appearance (Form)
Powder or solid
Titration Argentometric
>=96.0 to =<104.0 %
Appearance (Color)
White to off-white to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text