Thermo Scientific Chemicals

Diethyl carbonate, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436131000
100 mL, Each
Thermo Scientific Chemicals

Diethyl carbonate, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436131000
100 mL, Each
Quantity
Catalog number: 436131000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-58-8
IUPAC Name
diethyl carbonate
Molecular Formula
C5H10O3
InChI Key
OIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILES
CCOC(=O)OCC
Appearance (Form)
Liquid
Refractive index
1.3830 to 1.3850 (20°C, 589 nm)
Water
=<0.1 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text