Thermo Scientific Chemicals

n-Hexadecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436140010
1 L, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

n-Hexadecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436140010
1 L, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 436140010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
544-76-3
IUPAC Name
hexadecane
Molecular Formula
C16H34
InChI Key
DCAYPVUWAIABOU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC
Appearance (Form)
Clear liquid or solid
Infrared spectrum
Conforms
GC
>=98.5 %
Color scale
=<10 APHA
Refractive index
1.4330 to 1.4350 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text