Thermo Scientific Chemicals

Pentadecylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436160500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Pentadecylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436160500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 436160500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78887-70-4
IUPAC Name
magnesium(2+) pentadecan-1-ide bromide
Molecular Formula
C15H31BrMg
InChI Key
FAXYNDJJWMSBJG-UHFFFAOYSA-M
SMILES
[Mg++].[Br-].CCCCCCCCCCCCCC[CH2-]
Appearance (Color)
Yellow to brown
Appearance (Form)
Solution
Titration Grignard
15.0 to 17.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text