Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals
Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 436180050
Price (USD)/ Each
172.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
172.00
Add to cart
Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals
Catalog number436180050
Price (USD)/ Each
172.00
-
Add to cart
Chemical Identifiers
CAS72657-23-9
IUPAC Namemethyl 3-hydroxy-2-methylpropanoate
Molecular FormulaC5H10O3
InChI KeyATCCIZURPPEVIZ-UHFFFAOYNA-N
SMILESCOC(=O)C(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Specific optical rotation-23° to -29° (20°C, 589 nm) (c=4, methanol)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only