Thermo Scientific Chemicals

Pyridoxamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 436250010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyridoxamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 436250010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 436250010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
524-36-7
IUPAC Name
dihydrogen 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol dichloride
Molecular Formula
C8H14Cl2N2O2
InChI Key
HNWCOANXZNKMLR-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CC1=NC=C(CO)C(CN)=C1O
Titration Argentometric
>=97.5 to =<102.5 % (on dry basis)
Appearance (Color)
White to yellow to light brown
Infrared spectrum
Conforms
Appearance (Form)
Powder
Water
=<10 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text