1x1 image pixel for data collection
Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Thermo Scientific Chemicals

Pyridoxamine dihydrochloride, 98%

CAS: 524-36-7 | C8H12N2O2·2ClH | 241.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 436250050
Price (USD)
203.65
Special offer
Online exclusive
Ends: 26-Sep-2025
251.00
Save 47.35 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
203.65
Special offer
Online exclusive
Ends: 26-Sep-2025
251.00
Save 47.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS524-36-7
IUPAC Namedihydrogen 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol dichloride
Molecular FormulaC8H14Cl2N2O2
InChI KeyHNWCOANXZNKMLR-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CC1=NC=C(CO)C(CN)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 to =<102.5 % (on dry basis)
Appearance (Form)Powder
Water=<10 % (K.F.)
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
345201Certificate of AnalysisNov 23, 2023436250050, 436250010
343825Certificate of AnalysisDec 14, 2022436250050, 436250010
344846Certificate of AnalysisJun 10, 2022436250050, 436250010
344400Certificate of AnalysisSep 08, 2020436250050, 436250010
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection