Thermo Scientific Chemicals

2,4,6-Trimethylstyrene, 95%, Thermo Scientific Chemicals

Catalog number: 436280050
5 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylstyrene, 95%, Thermo Scientific Chemicals

Catalog number: 436280050
5 g, Each
Quantity
Catalog number: 436280050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
769-25-5
IUPAC Name
2-ethenyl-1,3,5-trimethylbenzene
Molecular Formula
C11H14
InChI Key
PDELBHCVXBSVPJ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C=C)C(C)=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94.0 %
Stabilizer
=<0.05 % tert-Butylcatechol

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text