Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific Chemicals
Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 436290050
Price (USD)/ Each
591.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
696.00 
Save 104.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
591.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
696.00 
Save 104.35 (15%)
Add to cart
Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific Chemicals
Catalog number436290050
Price (USD)/ Each
591.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
696.00 
Save 104.35 (15%)
-
Add to cart
Chemical Identifiers
CAS21085-72-3
IUPAC Namemethyl (2S,3S,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxane-2-carboxylate
Molecular FormulaC13H17BrO9
InChI KeyGWTNLHGTLIBHHZ-SVNGYHJRSA-N
SMILESCOC(=O)[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer2 % CaCO3
Appearance (Form)Crystalline powder or solid
HPLC>=97.5 %
Appearance (Color)White to grey
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only