Thermo Scientific Chemicals

3-Fluoro-4-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 436380050
5 g, Each
Thermo Scientific Chemicals

3-Fluoro-4-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 436380050
5 g, Each
Quantity
Catalog number: 436380050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22282-75-3
IUPAC Name
3-fluoro-4-iodopyridine
Molecular Formula
C5H3FIN
InChI Key
MSLUTDVOKZOXTA-UHFFFAOYSA-N
SMILES
FC1=CN=CC=C1I
Appearance (Color)
Cream to beige
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Crystalline powder
Melting point
87°C to 91°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text