Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 2 Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 2 x 5 mL |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 10 mL |
Unit Size | 2 x 5 mL |
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 1 Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 1 x 5 mL |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 5 mL |
Unit Size | 1 x 5 mL |
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 5 Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 5 x 5 mL |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 55 mL |
Unit Size | 5 x 5 mL |
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 10 Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 10 x 5 mL |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 50 mL |
Unit Size | 10 x 5 mL |
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 10 Unit Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 10 x 5 mL |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 50 mL |
Unit Size | 10 x 5 mL |
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Kit Type | 1 Bulk Pack |
Passive Reference Dye | ROX (Pre-mixed) |
Product Line | TaqMan™ |
Product Type | Universal PCR Master Mix |
Quantity | 1 x 50 mL |
Shipping Condition | Wet Ice |
For Use With (Application) | Gene Expression, Genotyping, miRNA analysis |
Sample Type | DNA (Genomic), cDNA |
Volume | 50 mL |
Unit Size | 1 x 50 mL |
Catalog Number | Specifications | Unit Size | Kit Type | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4364338 | 2 x 5 mL | 2 Pack | 1,255.65 Online price 1,342.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4304437 | 1 x 5 mL | 1 Pack | 656.65 Online price 712.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4364340 | 5 x 5 mL | 5 Pack | 2,955.65 Online price 3,155.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4305719 | 10 x 5 mL | 10 Pack | 5,626.65 Online price 6,150.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4318157 | 10 x 5 mL | 10 Unit Pack | 5,567.65 Online price 6,060.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4326708 | 1 x 50 mL | 1 Bulk Pack | 5,940.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
TaqMan Universal PCR Master Mix is the ideal reagent solution when you need a master mix for multiple 5' nuclease DNA applications. Applied Biosystems reagents have been validated with TaqMan assays and Applied Biosystems real-time systems to ensure sensitive, accurate, and reliable performance every time.
• Contains AmpliTaq Gold DNA Polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq DNA Polymerase
• Includes a passive internal reference based on proprietary ROX dye, for superb precision on Applied Biosystems real-time PCR instruments
• Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
• AmpErase UNG protects against subsequent re-amplification from PCR products containing dU to minimize carry-over contamination
• Rapid assay development guidelines are provided to minimize optimization time
Alternative product: Try TaqMan Fast Advanced Master Mix (Cat. No. 4444963), our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal PCR Master Mix and added additional capabilities for your gene expression analysis.