Applied Biosystems™

TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

Catalog number: 4364343
Have Questions?
Applied Biosystems™

TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

Catalog number: 4364343
Have Questions?
Catalog Number
4364343
Unit Size
1,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43643431,000 reactions
-
43240202,000 reactions
-
4324018200 reactions
-
43266142,000 reactions
-
4364341400 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
TaqMan Universal PCR Master Mix, No AmpErase UNG, is the ideal reagent solution when you need a master mix for multiple 5' nuclease DNA applications. Applied Biosystems reagents have been validated with TaqMan assays and Applied Biosystems real-time systems to ensure sensitive, accurate, and reliable performance every time.

• Contains AmpliTaq Gold DNA Polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq DNA Polymerase
• Includes a passive internal reference based on proprietary ROX dye, for superb precision on Applied Biosystems real-time PCR instruments
• Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
• Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase UNG
• Rapid assay development guidelines are provided to minimize optimization time

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal PCR Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye
ROX (Pre-mixed)
Product Line
TaqMan™
Product Type
Universal PCR Master Mix
Quantity
5 x 5 mL
Shipping Condition
Wet Ice
Sufficient For
1000 Reactions
For Use With (Application)
Gene Expression, Genotyping, miRNA analysis
No. of Reactions
1000 Reactions
Sample Type
DNA (Genomic), cDNA
Volume
55 mL

Contents & Storage

Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains 5 X 5 mL tubes, sufficient for 1000 reactions at the 50 μL total volume. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text