Thermo Scientific Chemicals

Trioctylphosphine, 90%, technical grade, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436461000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Trioctylphosphine, 90%, technical grade, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 436461000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 436461000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4731-53-7
IUPAC Name
trioctylphosphane
Molecular Formula
C24H51P
InChI Key
RMZAYIKUYWXQPB-UHFFFAOYSA-N
SMILES
CCCCCCCCP(CCCCCCCC)CCCCCCCC
Appearance (Color)
Clear to hazy colorless to yellow
Appearance
May develop some turbidity or precipitate
GC
>=88 %
Refractive index
1.4670 to 1.4690 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text