Thermo Scientific Chemicals

(S)-3-(BOC-Amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 436530010
1 g, Each
Thermo Scientific Chemicals

(S)-3-(BOC-Amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: 436530010
1 g, Each
Quantity
Catalog number: 436530010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
216854-23-8
IUPAC Name
tert-butyl N-(piperidin-3-yl)carbamate
Molecular Formula
C10H20N2O2
InChI Key
WUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC1CCCNC1
GC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Specific optical rotation
-2.5° to -3.5° (20°C, 589 nm) (c=1 in DMF)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text