Molecular sieves 4A, Cylinders, 3.2mm diameter, Thermo Scientific Chemicals
Molecular sieves 4A, Cylinders, 3.2mm diameter, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 4A, Cylinders, 3.2mm diameter, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number 436730050
Price (USD)/ Each
254.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
299.00 
Save 44.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
254.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
299.00 
Save 44.35 (15%)
Add to cart
Molecular sieves 4A, Cylinders, 3.2mm diameter, Thermo Scientific Chemicals
Catalog number436730050
Price (USD)/ Each
254.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
299.00 
Save 44.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
CAS70955-01-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Bulk density600 to 750 g/l
Residual water=<2 % (550°C, 2 h)
Appearance (Form)Cylinders
Water adsorption>=20 % (50% rH, 20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only