Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: 436761000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: 436761000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 436761000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
368-39-8
IUPAC Name
tetrafluoroboranuide; triethyloxidanium
Molecular Formula
C6H15BF4O
InChI Key
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CC[O+](CC)CC
Appearance (Form)
Powder
Titration with NaOH
>=96.0 %
Appearance (Color)
White to off-white
Stabilizer
3 to 5 % ether

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text