Applied Biosystems™

Power SYBR™ Green PCR Master Mix

Catalog number: 4368702
Have Questions?
Applied Biosystems™

Power SYBR™ Green PCR Master Mix

Catalog number: 4368702
Have Questions?
Catalog Number
4368702
Unit Size
5 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43687025 x 5 mL
-
43685771 x 1 mL
-
43676591 x 5 mL
-
43687062 x 5 mL
-
436870810 x 5 mL
-
43676601 x 50 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix. Power SYBR™ Master Mix offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.

• Everything you need for SYBR™ Green dye–based PCR amplification and detection in a convenient single-tube format. Power SYBR™ Green PCR Master Mix contains all of the components, excluding the template and primers, for superior SYBR™ Green reagent-based real-time PCR
• The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer greater sensitivity than classic SYBR™ Green PCR master mix
Power SYBR™ Green PCR Master Mix contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG, to minimize carryover contamination
• Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations to minimize well-to-well variability that result from a variety of causes, such as pipetting error and sample evaporation
Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions

Alternative product
Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Power SYBR Green PCR Master Mix and added additional capabilities for your gene expression analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Bottle
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD
Reaction Speed
Standard
For Use With (Equipment)
7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Label or Dye
SYBR Green
Passive Reference Dye
ROX (Pre-mixed)
Product Line
SYBR™
Product Type
Real Time PCR SYBR Master Mix
Purity or Quality Grade
LD (Low DNA)
Quantity
5 x 5 mL
Sufficient For
1000 Reactions of 50 μL Each
For Use With (Application)
Gene Expression
Concentration
2X
No. of Reactions
1000 Reactions
Sample Type
DNA (Genomic), cDNA
Volume
5 x 5 mL

Contents & Storage

The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Contains 1 × 50 mL bottle, sufficient for 1000 reactions of 50 μL each.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text