mono-Ethyl succinate, 95%, Thermo Scientific Chemicals
mono-Ethyl succinate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

mono-Ethyl succinate, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 436880050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1070-34-4
IUPAC Name4-ethoxy-4-oxobutanoic acid
Molecular FormulaC6H10O4
InChI KeyLOLKAJARZKDJTD-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Elemental Analysis46.4 to 52.3 % (C)
Elemental Analysis41.2 to 46.4 % (O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only