TaqMan™ Gene Expression Master Mix
Find Real-Time and Digital PCR technical tips, troubleshooting help, and resources within our new Real-Time PCR and Digital PCR Applications Support Center.
TaqMan™ Gene Expression Master Mix
Applied Biosystems™

TaqMan™ Gene Expression Master Mix

Get precise and reliable real-time qPCR quantification over nine logs of linear dynamic range with TaqMan Gene Expression Master Mix.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43690161 x 5 mL
436954210 x 5 mL
43700481 x 1 mL
43695142 x 5 mL
43695105 x 5 mL
43700741 x 50 mL
Catalog number 4369016
Price (USD)
605.65
Online Exclusive
668.00
Save 62.35 (9%)
Each
-
Add to cart
Quantity:
1 x 5 mL
Price (USD)
605.65
Online Exclusive
668.00
Save 62.35 (9%)
Each
Add to cart
Ask our AI about this Product
Get precise and reliable real-time qPCR quantification over nine logs of linear dynamic range with TaqMan Gene Expression Master Mix. TaqMan Gene Expression Master Mix is an optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target. Extended benchtop stability provides consistent results for automated liquid-handling systems, making it the choice for high-throughput sample processing, overnight or over the weekend. TaqMan Gene Expression Master Mix has been validated with TaqMan gene expression assays and Applied Biosystems real-time systems, providing precise quantification for a variety of gene expression–based real-time qPCR applications, including:
• Rare transcript detection
• Gene expression analysis
• Pathogen detection and viral load quantification
• Co-amplification of two targets
• Specific detection of gene family members

TaqMan Gene Expression Master Mix contains AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and uracil-DNA glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX dye for superb precision on Applied Biosystems real-time PCR instruments.

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Gene Expression Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR SystemStepOne™, Standard Mode, StepOnePlus™, Standard Mode
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan™
Purity or Quality GradeUP (Ultra Pure)
Quantity1 x 5 mL
Sample TypeDNA (Genomic), cDNA
Volume1 x 5 mL
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
FormLiquid
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
1 bottle (5 mL) 2X TaqMan™ Gene Expression Master Mix sufficient for 200 reactions. Store at 2°C to 8°C.


TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link