Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 436940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 436940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 436940250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-17-7
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Formula
H10Na2O8S2
InChI Key
PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration Iodimetric
>=98.5 %
Total trace metal impurities
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text