Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 436960250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS6484-52-2
IUPAC Nameammonium nitrate
Molecular FormulaH4N2O3
InChI KeyDVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES[NH4+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Total trace metal impurities=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only