Thermo Scientific Chemicals

Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 436970100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Manganese(II) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 436970100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 436970100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-43-0
IUPAC Name
oxomanganese
Molecular Formula
MnO
InChI Key
VASIZKWUTCETSD-UHFFFAOYSA-N
SMILES
O=[Mn]
Appearance (Form)
Crystalline powder and/or chunks
Appearance (Color)
Green
Titration Complexometric
76.7 to 78.2 % (Mn)
Total trace metal impurities
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text