Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 436980100
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
Add to cart
Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Catalog number436980100
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameindium(3+) triacetate
Molecular FormulaC6H9InO6
InChI KeyVBXWCGWXDOBUQZ-UHFFFAOYSA-K
SMILES[In+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol)291.95
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric35.3 to 43.3 % (In)
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only